missing translation for 'onlineSavingsMsg'
Learn More

OSTM1 Antibody (30), DyLight 550 , Novus Biologicals™

Product Code. 18430388 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430388 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430388 Supplier Novus Biologicals Supplier No. NBP289708R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

OSTM1 Monoclonal antibody specifically detects OSTM1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSTM1
Applications ELISA
Classification Monoclonal
Clone 30
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias GIPN, GLGAIP-interacting protein N terminus, grey-lethal osteopetrosis, HSPC019, OPTB5, osteopetrosis associated transmembrane protein 1, osteopetrosis-associated transmembrane protein 1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human OSTM1 extracellular domain (NP_054747.2; Met 1-Pro 284).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 28962
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.