missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTF1 Mouse anti-Human, Clone: 9G4, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59396-50UL
This item is not returnable.
View return policy
Description
OSTF1 Monoclonal specifically detects OSTF1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| OSTF1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1009, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| FLJ20559, OSFSH3P2bA235O14.1, osteoclast stimulating factor 1, osteoclast-stimulating factor 1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 9G4 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| OSTF1 | |
| Recombinant human OSTF1 (aa) purified from E. coli | |
| 50 μL | |
| Signal Transduction | |
| 26578 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction