missing translation for 'onlineSavingsMsg'
Learn More

OSTF1 Antibody (9G4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-59396MFV500

Product Code. 18453283

  • £433.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OSTF1 Monoclonal antibody specifically detects OSTF1 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OSTF1
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human
Purified
Flow Cytometry
9G4
Flow Cytometry
FLJ20559, OSFSH3P2bA235O14.1, osteoclast stimulating factor 1, osteoclast-stimulating factor 1
Recombinant human OSTF1 (1-217 aa) purified from E. coli
0.1 mL
Signal Transduction
26578
Store at 4C in the dark.
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.