missing translation for 'onlineSavingsMsg'
Learn More

Osterix/Sp7 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18388925 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388925 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388925 Supplier Novus Biologicals Supplier No. NBP310908100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Osterix/Sp7 Polyclonal specifically detects Osterix/Sp7 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osterix/Sp7
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry
Formulation PBS buffer, 2% sucrose
Gene Alias MGC126598, OSXosterix, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the C terminal region of human Osterix/Sp7 (NP_690599). Peptide sequence RTHGEPGPGPPPSGPKELGEGRSTGEEEASQTPRPSASPATPEKAPGGSP
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121340
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.