missing translation for 'onlineSavingsMsg'
Learn More

Osterix/Sp7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £484.00

Specifications

Antigen Osterix/Sp7
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18699905
View Documents
Novus Biologicals
NBP2-38019-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18161019
View Documents
Novus Biologicals
NBP2-38019
0.1 mL
£484.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Osterix/Sp7 Polyclonal specifically detects Osterix/Sp7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Osterix/Sp7
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
MGC126598, OSXosterix, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix
SP7
IgG
Affinity Purified
Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Polyclonal
Rabbit
Human
Q8TDD2
121340
This antibody was developed against a recombinant protein corresponding to amino acids: QGQGDGLQGTLPTGPAQPPLNPQLPTYPSDFAPLNPAPYPAPHLLQPGPQHVLPQDVYKPKAVGNSG
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.