missing translation for 'onlineSavingsMsg'
Learn More

Osteoprotegerin/TNFRSF11B Antibody (69127) [DyLight 594], Novus Biologicals™

Product Code. 30733792 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733792 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733792 Supplier Novus Biologicals Supplier No. FAB8051M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Osteoprotegerin/TNFRSF11B Monoclonal antibody specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Applications ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 69127
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Osteoprotegerin/TNFRSF11B, Glu22-Leu401, Accession # O00300
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 4982
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.