missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteoprotegerin/TNFRSF11B Antibody (340), mFluor Violet 500 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Osteoprotegerin/TNFRSF11B Monoclonal antibody specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | Osteoprotegerin/TNFRSF11B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 340 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human Osteoprotegerin/TNFRSF11B (Accession#: NP_002537.3; Met 1-Leu 401). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?