missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteoprotegerin/TNFRSF11B Antibody (340), Alexa Fluor™ 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89371AF350
This item is not returnable.
View return policy
Description
Osteoprotegerin/TNFRSF11B Monoclonal antibody specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for ELISA
Specifications
| Osteoprotegerin/TNFRSF11B | |
| Monoclonal | |
| Alexa Fluor 350 | |
| OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TR1, tumor necrosis factor receptor superfamily, member 11b | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Osteoprotegerin/TNFRSF11B (NP_002537.3; Met 1-Leu 401). | |
| 0.1 mL | |
| Apoptosis, Biologically Active Proteins, Cell Biology | |
| 4982 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 340 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction