Learn More
Osteopontin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 500-P314BT-1MG
Description
AA Sequence of recombinant protein: IPVKQADSGS SEEKQLYNKY PDAVATWLNP DPSQKQNLLA PQNAVSSEET NDFKQETLPS KSNESHDHMD DMDDEDDDDH VDSQDSIDSN DSDDVDDTDD SHQSDESHHS DESDELVTDF PTDLPATEVF TPVVPTVDTY DGRGDSVVYG LRSKSKKFRR PDIQYPDATD EDITSHMESE ELNGAYKAIP VAQDLNAPSD WDSRGKDSYE TSQLDDQSAE THSHKQSRLY KRKANDESNE HSDVIDSQEL SKVSREFHSH EFHSHEDMLV VDPKSKEEDK HLKFRISHEL DSASSEVN. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Osteopontin. Anti-Human Osteopontin-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human Osteopontin by sandwich ELISA (using 100 mL/well), a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti- Human Osteopontin (500-P314) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Osteopontin. Western Blot: To detect Human Osteopontin by Western Blot analysis, this antibody can be used at a concentration of 0.1-0.2 mg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Human Osteopontin is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P314BT-1MG will be provided as 2 x 500 μg
Osteopontin is an arginine-glycine-aspartic acid (RGD)-containing glycoprotein that interacts with integrins and CD44 as major receptors. It is a multifunctional protein involved in bone mineralization, cell adhesion, cell migration, chronic inflammatory disease and transformation. Proteolytic cleavage by thrombin and matrix metalloproteinases close to the integrin-binding Arg-Gly-Asp sequence modulates the function of OPN and its integrin binding properties. Thrombin-cleaved fragments of Osteopontin are overexpressed in malignant glial tumors and provide a molecular niche with survival advantage and provide a novel substrate for plasmin and cathepsin D.
Specifications
| Osteopontin | |
| Polyclonal | |
| Biotin | |
| SPP1 | |
| 2AR; 2b7; 44 kDa bone phosphoprotein; Apl-1; BNSP; Bone sialoprotein 1; Bone sialoprotein 1 (BSPI/BSP1); Bsp; BSPI; Calcium oxalate crystal growth inhibitor protein; CALPHA1 fusion; Early T lymphocyte activation 1 (ETA1); early T-lymphocyte activation 1; early T-lymphocyte activation 1 protein; Eta; ETA-1; HGNC:11255; immunoglobulin alpha 1 heavy chain constant region fusion protein; MGC110940; Minopontin; nephropontin; Op; Opn; Opnl; OSP; osteopontin; osteopontin/immunoglobulin alpha 1 heavy chain constant region fusion protein; osteopontin-like protein; PSEC0156; Ric; Secreted phosphoprotein 1; secreted phosphoprotein 1 (osteopontin bone sialoprotein I early T lymphocyte activation 1); secreted phosphoprotein 1 (osteopontin, bone sialoprotein I, early T-lymphocyte activation 1); Secreted phosphoprotein 1 (SPP1); secreted phosphoprotein 1 variant 6; Sialoprotein (osteopontin); SPP1; SPP-1; SPP1/CALPHA1 fusion; unnamed protein product; urinar; urinary stone protein; Uropontin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6696 | |
| -20°C |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P10451 | |
| SPP1 | |
| E.coli-derived, 33.7 kDa Recombinant Human Osteopontin | |
| 2 x 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.