missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Osteopontin/OPN Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
£307.00 - £448.00
Specifications
| Antigen | Osteopontin/OPN |
|---|---|
| Clone | 1 |
| Dilution | Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204538
|
Novus Biologicals
NBP2-89443-50UL |
50 μg |
£307.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263607
|
Novus Biologicals
NBP2-89443-100UL |
100 μg |
£448.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Osteopontin/OPN | |
| Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
| Recombinant Human Osteopontin/OPN protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Extracellular Matrix, Hypoxia | |
| 0.2 um filtered solution in PBS | |
| 6696 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title