missing translation for 'onlineSavingsMsg'
Learn More

Osteopontin/OPN Antibody (223112) [DyLight 594], Novus Biologicals™

Product Code. 30736060 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30736060 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736060 Supplier Novus Biologicals Supplier No. FAB14332M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Osteopontin/OPN Monoclonal antibody specifically detects Osteopontin/OPN in Human samples. It is validated for ELISA,ELISA Standard (Matched Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteopontin/OPN
Applications ELISA, ELISA Standard (Matched Pair)
Classification Monoclonal
Clone 223112
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias BNSP, Bone sialoprotein 1, MGC110940, Nephropontin, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Osteopontin, Ile17-Asn300, Accession # NP_000573.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cancer, Cellular Markers, Extracellular Matrix, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 6696
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.