missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Osteocrin Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148622
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Hormone that acts as a regulator of dendritic growth in the developing cerebral cortex in response to sensory experience. Induced in the brain following membrane depolarization and inhibits dendritic branching in neurons of the developing cortex.
Specifications
| Osteocrin | |
| Polyclonal | |
| Unconjugated | |
| OSTN | |
| MUSCLIN; Ostc; osteocrin; Ostn; Processed Osteocrin | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 239790, 344901 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P61364, P61366 | |
| OSTN | |
| E. coli-derived recombinant human Osteocrin, Val28-Gly133. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction