missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Osteocalcin Monoclonal Antibody (OCG4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA120788
This item is not returnable.
View return policy
Description
This antibody does not detect rat or mouse Osteocalcin. Predicted molecular weight: 12kDa.
Osteocalcin belongs to the osteocalcin/matrix Gla-protein family and constitutes 1-2% of the total bone protein. It binds strongly to apatite and calcium. Gamma-carboxyglutamate residues are formed by vitamin K dependent carboxylation. These residues are essential for the binding of calcium.
Specifications
| Osteocalcin | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 1% BSA and no preservative; pH 7.4 | |
| P02818, P02820, P02822, P0C225, P39056, Q8HYY9 | |
| BGLAP | |
| Bovine osteocalcin. | |
| 50 μg | |
| Primary | |
| Human, Rabbit, Bovine, Sheep, Pig, Chicken, Goat | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| OCG4 | |
| Unconjugated | |
| BGLAP | |
| Bglap; Bglap1; Bglap2; BGP; Bgpr; Bgpra; bone gamma carboxyglutamate protein; bone gamma carboxyglutamate protein 1; bone gamma-carboxyglutamate (gla) protein; bone gamma-carboxyglutamate (gla) protein (osteocalcin); bone gamma-carboxyglutamate protein; bone gamma-carboxyglutamate protein 2; bone gamma-carboxyglutamate protein osteocalcin; bone gamma-carboxyglutamate; BGLAP; bone gamma-carboxyglutamic acid (Gla) protein (osteocalcin); bone Gla protein; bone gla protein BGP; bone Gla-protein; gamma-carbooxyglutamate (gla) protein; Gamma-carboxyglutamic acid-containing protein; GLA; Gla protein precusor; mOC-A; OC; OCN; OG1; Osteocalcin; PMF1; RP11-54H19.5; unnamed protein product | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100340503, 100861188, 281646, 396348, 397530, 632, 677670 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction