missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSTC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595894
This item is not returnable.
View return policy
Description
OSTC Polyclonal Antibody for Western Blot, ELISA
OSTC is a protein coding gene.
Specifications
| OSTC | |
| Polyclonal | |
| Unconjugated | |
| OSTC | |
| 2310008M10Rik; 5730557H03Rik; DC2; HDCMD45P; HSPC307; Hydrophobic protein HSF-28; oligosaccharyltransferase complex non-catalytic subunit; oligosaccharyltransferase complex subunit; oligosaccharyltransferase complex subunit (non-catalytic); oligosaccharyltransferase complex subunit OSTC; Ostc; ostc {ECO:0000250; RGD1560708; UniProtKB:Q9NRP0} | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 362040, 58505, 66357 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.89 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| B0K025, Q78XF5, Q9NRP0 | |
| OSTC | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction