missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSR2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116805
This item is not returnable.
View return policy
Description
Positive Samples: SKOV3, 293T, Mouse testis, Mouse pancreas, Rat large intestine, Rat kidney Immunogen sequence: HMQESPHKCP TCGRTFNQRS NLKTHLLTHT DIKPYSCEQC GKVFRRNCDL RRHSLTHTPR QDF.
OSR2 is a mammalian homolog of the Drosophila odd-skipped family of transcription factors.
Specifications
| OSR2 | |
| Polyclonal | |
| Unconjugated | |
| OSR2 | |
| 5430409I15Rik; odd-skipped related 2; odd-skipped related transciption factor 2; Osr2; Osr2A; Osr2B; Protein odd-skipped-related 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 107587, 116039, 315039 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.91 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6AY34, Q8N2R0, Q91ZD1 | |
| OSR2 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-312 of human OSR2 (NP_0011359341). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction