missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116814
This item is not returnable.
View return policy
Description
Positive Samples: NCI-H460, Cal-27, A-549, 22Rv1. Immunogen sequence: NGLPTVPSDH LPNLYGFSAL HAVHLHQWTL GYPAMHLPRS SFSKVPGTVS SLVDARFQLP AFPWFPHVIQ PKPEITAGGS VPALKTKP.
OSR1 is a transcription factor that plays a role in the regulation of embryonic heart and urogenital development.
Specifications
| OSR1 | |
| Polyclonal | |
| Unconjugated | |
| Osr1 | |
| ODD; Odd1; odd-skipped homolog; odd-skipped related 1; odd-skipped related 1 (Drosophila); odd-skipped related transciption factor 1; odd-skipped related transcription factor 1; Osr; OSR1; Protein odd-skipped-related 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 130497, 23967, 298878 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.68 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| B0K011, Q8TAX0, Q9WVG7 | |
| Osr1 | |
| Recombinant protein (or fragment)'. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction