missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSMR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA593143
This item is not returnable.
View return policy
Description
Positive Samples: 293T, HeLa, HepG2, Raji, Mouse lung, Mouse heart; Cellular Location: Membrane, Single-pass type I membrane protein Immunogen sequence: ESELPLECAT HFVRIKSLVD DAKFPEPNFW SNWSSWEEVS VQDSTGQDIL FVFPKDKLVE EGTNVTICYV SRNIQNNVSC YLEGKQIHGE QLDPHVTAFN L.
Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF.Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
| OSMR | |
| Polyclonal | |
| Unconjugated | |
| OSMR | |
| IL-31 receptor subunit beta; IL-31R subunit beta; IL-31RB; IL-31R-beta; interleukin-31 receptor subunit beta; MGC150626; MGC150627; MGC75127; oncostatin M receptor; oncostatin receptor; oncostatin-M specific receptor beta subunit; oncostatin-M specific receptor subunit beta; oncostatin-M-specific receptor subunit beta; OSMR; Osmrb; PLCA1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18414, 9180 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.62 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O70458, Q99650 | |
| OSMR | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction