missing translation for 'onlineSavingsMsg'
Learn More

OSMR beta Antibody (002), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18406229 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406229 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406229 Supplier Novus Biologicals Supplier No. NBP289850MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

OSMR beta Monoclonal antibody specifically detects OSMR beta in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSMR beta
Applications Flow Cytometry
Classification Monoclonal
Clone 2
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human OSMR beta (NP_003990.1; Met1-Met740).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9180
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.