missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSMR beta Antibody (002), Alexa Fluor™ 594, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89850AF594
This item is not returnable.
View return policy
Description
OSMR beta Monoclonal specifically detects OSMR beta in Human samples. It is validated for Flow Cytometry.
Specifications
| OSMR beta | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 9180 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 2 | |
| Flow Cytometry | |
| IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human OSMR beta (NP_003990.1; Met1-Met740). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction