missing translation for 'onlineSavingsMsg'
Learn More

Osmium(VIII) oxide, 99.8% (metals basis), Os 74.4% min

Product Code. 11356439
Change view
Click to view available options
Quantity:
0.5 g
1 g
10 x 1 g
Unit Size:
0.5g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356439 1 g 1g
11346439 0.5 g 0.5g
11366439 10 x 1 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356439 Supplier Thermo Scientific Alfa Aesar Supplier No. 012103.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20816-12-0 | O4Os | 254.23 g/mol

Oxidation, catalyst in Sharpless dihydroxylations Osmium(VIII) oxide is used in the Sharpless oxyamination to give vicinal amino-alcohols. It is used to oxidize alkenes to the vicinal diols. It acts as a staining agent used in transmission electron microscopy to provide contrast to the image and as a stain for lipids in optical microscopy. As a polymer stain, it is also used to stain selectively copolymers especially styrene-butadiene block copolymers. Further, it is involved in the synthesis of Isosteviol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Boiling Point 130°C
Melting Point 40.6°C
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 99.8% (metals basis)
Solubility Information Soluble in chloroform,alcohol and ethers.Soluble in water,organic solvents,benzene,alcohol,ether,ammonium hydroxide,phosphorus oxychloride and carbon tetrachloride.
Formula Weight 254.1
Odor Like chlorine
Appearance Crystalline
Vapor Pressure 10 hPa (8mm Hg) at 20°C
Chemical Name or Material Osmium(VIII) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.