missing translation for 'onlineSavingsMsg'
Learn More

Osmium Tetroxide Solution, Honeywell™ Fluka™

Product Code. 15678340 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass Bottle
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15678340 10 mL Glass Bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15678340 Supplier Honeywell-Fluka Supplier No. 25175510ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 20816-12-0
Quantity 10 mL
Molecular Formula O4Os
Linear Formula OsO4
MDL Number MFCD00011150
UN Number UN3287
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
SMILES O=[Os](=O)(=O)=O
IUPAC Name tetraoxoosmium
Molecular Weight (g/mol) 254.23
PubChem CID 30318
Formula Weight 254.23g/mol
Percent Purity 0%
Packaging Glass Bottle
Vapor Pressure 3.55psi (55°C), 1.23psi (20°C)
Density 1.0g/mL at 20°C, 1.04g/mL at 25°C
Chemical Name or Material Osmium Tetroxide Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.