missing translation for 'onlineSavingsMsg'
Learn More

Osmium Tetroxide, Honeywell™

Product Code. 15601730 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15601730 100 mg 100mg
15621730 250 mg 250mg
15611730 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15601730 Supplier Honeywell Research Chemicals Supplier No. 201030100MG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, 99.8%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20816-12-0
Molecular Formula O4Os
Molecular Weight (g/mol) 254.23
MDL Number MFCD00011150
InChI Key VUVGYHUDAICLFK-UHFFFAOYSA-N
Synonym osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4
PubChem CID 30318
IUPAC Name tetraoxoosmium
SMILES O=[Os](=O)(=O)=O

Specifications

Quantity 100 mg
Linear Formula OsO4
UN Number 2471
Formula Weight 254.23g/mol
Percent Purity 99%
Grade ReagentPlus
Vapor Pressure 7mmHg (20°C)
Density Vapor Density: 8.8 (vs air)
Chemical Name or Material Osmium Tetroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.