missing translation for 'onlineSavingsMsg'
Learn More

Osmium powder, -200 mesh, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog Number 11391799
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
7440-04-2
Os
190.23
MFCD00011147
SYQBFIAQOQZEGI-UHFFFAOYSA-N
ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
23937
CHEBI:30687
osmium
[Os]
This item is not returnable. View return policy
7440-04-2
Os
190.23
MFCD00011147
SYQBFIAQOQZEGI-UHFFFAOYSA-N
ion, metallic, osmium, elemental, unii-2e7m255opy, os4, osmio, hydrido, hydride, atom, powder
23937
CHEBI:30687
osmium
[Os]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
UN Number UN3089
Quantity 5 g
Percent Purity 99.8%
Mesh Size −200 Mesh
Odor Strong
Form Powder
Assay (metals basis)
Chemical Name or Material Osmium powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'