missing translation for 'onlineSavingsMsg'
Learn More

Osmium powder, -20 mesh, 99.95% (metals basis)

Product Code. 11359197
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359197 5 g 5g
11369197 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359197 Supplier Thermo Scientific Alfa Aesar Supplier No. 039998.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-04-2 | Os | 190.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4-2-7440
PubChem CID 23937
ChEBI CHEBI:30687
IUPAC Name osmium

Specifications

Quantity 5 g
Mesh Size -20 Mesh
Assay Percent Range 99.95% (metals basis)
Molecular Formula Pd
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES [Pd]
Molecular Weight (g/mol) 106.42
Chemical Name or Material Osmium powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.