missing translation for 'onlineSavingsMsg'
Learn More

Osmium, plasma standard solution, Specpure™ Os 1000μg/mL, Thermo Scientific Chemicals

Catalog Number 11361449
Click to view available options
Quantity:
50 mL
100 mL
500 mL
MFCD00011147
MFCD00011147

Osmium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration (NH4)2OsCl6 in 20% HCl
Quantity 50 mL
UN Number UN1789
Grade Specpure™
Solubility Miscible with water
Physical Form Liquid
Chemical Name or Material Osmium plasma standard solution, Os 1000μg/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'