missing translation for 'onlineSavingsMsg'
Learn More

Osmium(IV) oxide, Os 83% min

Product Code. 11368298
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368298 5 g 5g
11388298 0.25 g 0.25g
11378298 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11368298 Supplier Thermo Scientific Alfa Aesar Supplier No. 039497.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12036-02-1 | O2Os | 222.228 g/mol

The osmium oxide/hexacyanoruthenate films modified electrodes plays an important role for the electrocatalytic reaction of catecholamines and sulfur oxoanions. Used as catalyst. Metallic osmium or an osmium oxide-ruthenium cyanide film modified graphite electrodes plays an essential role for the determination of biogenic amines from electrocatalytic responses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-02-1
Molecular Formula O2Os
Molecular Weight (g/mol) 222.228
MDL Number MFCD00011150
InChI Key XSXHWVKGUXMUQE-UHFFFAOYSA-N
Synonym osmium iv oxide, osmium dioxide, osmium oxide oso2 6ci,7ci,8ci,9ci, osmium iv-oxid, acmc-1btxg, osmium iv oxide, os
PubChem CID 187574
IUPAC Name dioxoosmium
SMILES O=[Os]=O

Specifications

Color Black
Physical Form Solid
Quantity 5 g
Solubility Information Slightly soluble in cold water.
Formula Weight 222.2
Appearance Black powder
Density 11.4
Chemical Name or Material Osmium(IV) oxide
compliance-icons
Product Identifier
  • Osmium(IV) oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Osmium(IV) oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.