missing translation for 'onlineSavingsMsg'
Learn More

Osilodrostat, MedChemExpress

Product Code. 16477018
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477018

Brand: Medchem Express HY16276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Osilodrostat (LCI699) is a potent, orally active11β-hydroxylase (CYP11B1) inhibitor with an IC50 value of 35 nM.

Osilodrostat is a potent, orally aldosterone synthase (CYP11B2) inhibitor with IC50 values of 0.7 nM and 160 nM for human aldosterone synthase and rat aldosterone synthase, respectively. Osilodrostat inhibits aldosterone and corticosterone synthesis. Osilodrostat has blood pressure lowering ability. Osilodrostat can be used for research of Cushing syndrome (CS).

Specifications

Chemical Name or Material Osilodrostat
CAS 928134-65-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C13H10FN3
Quantity 10 mg
Synonym LCI699
Solubility Information DMSO : 250 mg/mL (1100.16 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (440.06 mM; Need ultrasonic)
SMILES N#CC1=CC=C([C@H]2CCC3=CN=CN32)C(F)=C1
Molecular Weight (g/mol) 227.24
Formula Weight 227.24
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.