missing translation for 'onlineSavingsMsg'
Learn More

OSI-027, MedChemExpress

Product Code. 16496677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496677 5 mg 5mg
16470317 50 mg 50mg
16413968 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496677 Supplier Medchem Express Supplier No. HY10423
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress OSI-027 (ASP7486) is a potent, selective, orally active and ATP-competitive mTOR kinase activity inhibitor with an IC50 of 4 nM. OSI-027 targets both mTORC1 and mTORC2 with IC50s of 22 nM and 65 nM, respectively.

OSI-027 (ASP7486) is a potent, selective, orally active and ATP-competitive mTOR kinase activity inhibitor with an IC50 of 4 nM. OSI-027 targets both mTORC1 and mTORC2 with IC50s of 22 nM and 65 nM, respectively.

Specifications

Chemical Name or Material OSI-027
CAS 936890-98-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H22N6O3
Quantity 5 mg
Synonym ASP7486
Solubility Information DMSO : 125 mg/mL (307.55 mM; Need ultrasonic)
SMILES O=C([C@H]1CC[C@H](C2=NC(C(N3)=CC4=C3C(OC)=CC=C4)=C5C(N)=NC=NN52)CC1)O
Molecular Weight (g/mol) 406.44
Formula Weight 406.44
Percent Purity 95.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.