missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSGIN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570467
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 87%; Dog: 100%; Guinea Pig: 80%; Horse: 100%; Human: 100%; Mouse: 87%; Pig: 93%; Rabbit: 100%; Rat: 93%.
May be involved in meiosis or the maturation of germ cells.
Specifications
| OSGIN2 | |
| Polyclonal | |
| Unconjugated | |
| OSGIN2 | |
| BC010311; C230027H09Rik; C8orf1; hT41; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 2; OSGIN2; oxidative stress induced growth inhibitor family member 2; oxidative stress-induced growth inhibitor 2; RGD1306323 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 734 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y236 | |
| OSGIN2 | |
| synthetic peptide directed towards the C terminal region of human OSGIN2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction