missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGIN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94192-0.02ml
This item is not returnable.
View return policy
Description
OSGIN1 Polyclonal antibody specifically detects OSGIN1 in Human samples. It is validated for Western Blot
Specifications
| OSGIN1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| BDGI, bone marrow stromal cell-derived growth inhibitor, EC 1.6.5, EC 2.4.2.7, EC 2.7.1.130, huOKL38, OKL38oxidative stress-induced growth inhibitor 1, Ovary, kidney and liver protein 38, oxidative stress induced growth inhibitor 1, pregnancy induced growth inhibitor, Pregnancy-induced growth inhibitor OKL38 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human OSGIN1 (NP_892026.1). MSSSRKDHLGASSSEPLPVIIVGNGPSGICLSYLLSGYTPYTKPDAIHPHPLLQRKLTEAPGVSILDQDLDYLSEGLEGRSQSPVALLFDALLRPDTDFGGNMKSVLTWKHRKEHAIPHVVLGRNLPGGAWHSIEGSMVILSQGQWMGLPDLEVKDWMQKKRRGLRNSRATAGDIAHYYRDYVVKKGLGHNFVSGAVVTA | |
| 0.02 mL | |
| Cell Biology | |
| 29948 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction