missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSGEP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558914
This item is not returnable.
View return policy
Description
Immunogen sequence: FEGSANKIGV GVVRDGKVLA NPRRTYVTPP GTGFLPGDTA RHHRAVILDL LQEALTESGL TSQDIDCIAY TKGP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Osgep (O-sialoglycoprotein endopeptidase), also known as GCPL1, is a 335 amino acid protein that is a member of the peptidase M22 family. Osgep specifically cleaves the 31-Arg-l-Asp-32 bond in glycophorin A, but it does not cleave desialylated glycoproteins, unglycosylated proteins or glycoproteins that are only N-glycosylated. Though ubiquitously expressed at low levels, highest levels of Osgep are found in liver, skeletal muscle and kidney. OSGEPL1, also known as Qri7, is a 414 amino acid protein that belongs to the KAE1/YgjD family and exists as three alternatively spliced isoforms. In tRNAs that have codons beginning with adenine, OSGEPL1 is required for the formation of a threonylcarbamoyl group on adenosine.
Specifications
| OSGEP | |
| Polyclonal | |
| Unconjugated | |
| OSGEP | |
| 1500019L24Rik; GCPL1; GCPL-1; HAMAP-Rule:MF_03180}; hOSGEP; KAE1; mOsgep; N6-L-threonylcarbamoyladenine synthase; OSGEP; OSGEP1; O-sialoglycoprotease; O-sialoglycoprotein endopeptidase; O-sialoglycoprotein endopeptidase {ECO:0000255; pasteurella haemolytica metalloprotease homolog with glycoprotein substrates/gpc-like protein 1; probable O-sialoglycoprotein endopeptidase; probable tRNA N6-adenosine threonylcarbamoyltransferase; probable tRNA N6-adenosine threonylcarbamoyltransferase {ECO:0000255; probable tRNA threonylcarbamoyladenosine biosynthesis protein Osgep; PRSMG1; Prsmg1/Gcpl1; t(6)A synthase; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP; t(6)A37 threonylcarbamoyladenosine biosynthesis protein Osgep {ECO:0000255; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEP; tRNA threonylcarbamoyladenosine biosynthesis protein Osgep {ECO:0000255 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55644 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPF4 | |
| OSGEP | |
| Recombinant protein corresponding to Human OSGEP. Recombinant protein control fragment (Product #RP-98345). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction