missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSGEP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £364.00
Specifications
| Antigen | OSGEP |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
OSGEP Polyclonal antibody specifically detects OSGEP in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| OSGEP | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 55644 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.4.24.57, GCPL1FLJ20411, hOSGEP, KAE1, OSGEP1, O-sialoglycoprotein endopeptidase, probable O-sialoglycoprotein endopeptidase, PRSMG1, t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-335 of human OSGEP (NP_060277.1). MPAVLGFEGSANKIGVGVVRDGKVLANPRRTYVTPPGTGFLPGDTARHHRAVILDLLQEALTESGLTSQDIDCIAYTKGPGMGAPLVSVAVVARTVAQLWNKPLVGVNHCIGHIEMGRLITGATSPTVLYVSGGNTQVIAYSEHRYRIFGETIDIAVGNCLDRFARVLKISNDPSPGYNIEQMAKRGKKLVELPYTVKGMDVSFSGILSFIEDVAHRMLATGECTPEDLCFSLQETVFAMLVEITERAMAHCGSQEALIVGGVGCNVRLQEMMATMCQERGARLFATDERFCIDNGAMIAQAGWEMFRAGHRTPLSDSGVTQRYRTDEVEVTWRD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title