missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSER1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560933
This item is not returnable.
View return policy
Description
Immunogen sequence: GKPCTCIGKE CQCKRWHDME VYSFSGLQSV PPLAPERRST LEDYSQSLHA RTLSGSPRSC SEQARVFVDD VTIEDLSGYM EYYLYIPKKM S Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.
Specifications
| OSER1 | |
| Polyclonal | |
| Unconjugated | |
| OSER1 | |
| 3230401D17Rik; AI256769; BM-038; C20orf111; dJ1183I21.1; HSPC168; HSPC207; OSER1; Osr1; oxidative stress responsive 1; oxidative stress responsive serine rich 1; oxidative stress responsive serine-rich 1; oxidative stress-responsive 1; oxidative stress-responsive protein 1; Oxidative stress-responsive serine-rich protein 1; Perit1; Peroxide-inducible transcript 1 protein; RGD1303142 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NX31 | |
| OSER1 | |
| Recombinant protein corresponding to Human OSER1. Recombinant protein control fragment (Product #RP-99434). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction