missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSER1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113346
This item is not returnable.
View return policy
Description
OSER1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Specifications
| OSER1 | |
| Polyclonal | |
| Unconjugated | |
| OSER1 | |
| 3230401D17Rik; AI256769; BM-038; C20orf111; dJ1183I21.1; HSPC168; HSPC207; OSER1; Osr1; oxidative stress responsive 1; oxidative stress responsive serine rich 1; oxidative stress responsive serine-rich 1; oxidative stress-responsive 1; oxidative stress-responsive protein 1; Oxidative stress-responsive serine-rich protein 1; Perit1; Peroxide-inducible transcript 1 protein; RGD1303142 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 51526, 66680 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9D722, Q9NX31 | |
| OSER1 | |
| Recombinant Human Oxidative stress-responsive serine-rich protein 1 protein (1-292AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction