missing translation for 'onlineSavingsMsg'
Learn More

Oseltamivir phosphate, MedChemExpress

Product Code. 16451826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451826 500 mg 500mg
16407236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451826 Supplier Medchem Express Supplier No. HY17016
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Oseltamivir phosphate (GS 4104) is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B.

Oseltamivir phosphate (GS 4104) is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B.

Specifications

Chemical Name or Material Oseltamivir phosphate
CAS 204255-11-8
Color White
Purity Grade Notes Research
Molecular Formula C16H31N2O8P
Quantity 500 mg
Synonym GS 4104
Solubility Information H2O : 100 mg/mL (243.66 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (243.66 mM; Need ultrasonic)
SMILES O=C(OCC)C1=C[C@H]([C@@H]([C@H](C1)N)NC(C)=O)OC(CC)CC.OP(O)(O)=O
Molecular Weight (g/mol) 410.4
Formula Weight 410.4
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.