missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Oseltamivir phosphate
GREENER_CHOICE

Product Code. 18735506
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18735506 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18735506 Supplier Tocris Bioscience™ Supplier No. 7224/100
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Influenza virus neuraminidase inhibitor; antiviral prodrug

Oseltamivir phosphate is a viral neuraminidase inhibitor prodrug; metabolized to GS 4071. Metabolite exhibits potent inhibition of neuraminidase from multiple influenza strains (IC50 values range from 0.3 to 2 nM). Inhibits cellular effects of influenza infection in vitro (IC50 values range from 0.6 to 150 nM). Displays highest potency against H1N1 and H3N2 influenza A strains. Inhibits H1N1 influenza infection in mice.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl (3 R,4 R,5 S)-4-Acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-ene-1-carboxylate phosphate
CAS 204255-11-8
Quantity 100 mg
Target Hydrolase Inhibitors
Molecular Formula C16H28N2O4.H3PO4
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.