missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oseltamivir acid, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Oseltamivir acid (GS 4071), the active metabolite of Oseltamivir phosphate, is an orally bioavailable, potent and selective inhibitor of influenza virus neuraminidase (IC50=2 nM) with activity against both influenza A and B viruses.
Specifications
Specifications
| Chemical Name or Material | Oseltamivir acid |
| CAS | 187227-45-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H24N2O4 |
| Quantity | 5 mg |
| Synonym | GS 4071 Ro 64-0802 Oseltamivir carboxylate |
| Solubility Information | DMSO : ≥ 230 mg/mL (808.86 mM) ∣H2O : 125 mg/mL (439.60 mM; Need ultrasonic) |
| SMILES | O=C(O)C1=C[C@H]([C@@H]([C@H](C1)N)NC(C)=O)OC(CC)CC |
| Molecular Weight (g/mol) | 284.35 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction