missing translation for 'onlineSavingsMsg'
Learn More

Oseltamivir acid, MedChemExpress

Product Code. 16485638
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485638 10 mg 10mg
16448337 5 mg 5mg
16471807 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485638 Supplier Medchem Express Supplier No. HY13318
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Oseltamivir acid (GS 4071), the active metabolite of Oseltamivir phosphate, is an orally bioavailable, potent and selective inhibitor of influenza virus neuraminidase (IC50=2 nM) with activity against both influenza A and B viruses.

Oseltamivir acid (GS 4071), the active metabolite of Oseltamivir phosphate, is an orally bioavailable, potent and selective inhibitor of influenza virus neuraminidase (IC50=2 nM) with activity against both influenza A and B viruses.

Specifications

Chemical Name or Material Oseltamivir acid
CAS 187227-45-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H24N2O4
Quantity 10 mg
Synonym GS 4071 Ro 64-0802 Oseltamivir carboxylate
Solubility Information DMSO : ≥ 230 mg/mL (808.86 mM) ∣H2O : 125 mg/mL (439.60 mM; Need ultrasonic)
SMILES O=C(O)C1=C[C@H]([C@@H]([C@H](C1)N)NC(C)=O)OC(CC)CC
Molecular Weight (g/mol) 284.35
Formula Weight 284.35
Percent Purity 95.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.