missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSCAR Antibody (RM0300-11E34), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
OSCAR Monoclonal antibody specifically detects OSCAR in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | OSCAR |
| Applications | Western Blot |
| Classification | Monoclonal |
| Host Species | Rat |
| Immunogen | Mouse recombinant OSCAR (extracellular domain) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?