missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ OSBPL9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29894
This item is not returnable.
View return policy
Description
OSBPL9 Polyclonal antibody specifically detects OSBPL9 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| OSBPL9 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| FLJ12492, FLJ14629, FLJ14801, FLJ32055, FLJ34384, ORP-9, ORP9MGC15035, OSBP4, OSBP-related protein 9, oxysterol binding protein-like 9, oxysterol-binding protein-related protein 9 | |
| Between 686 and 736 | |
| 100 μg | |
| Lipid and Metabolism | |
| 114883 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction