missing translation for 'onlineSavingsMsg'
Learn More

OSBPL5 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NB100-57071

Product Code. 18009625

  • £427.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

OSBPL5 Polyclonal specifically detects OSBPL5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OSBPL5
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
EC 1.2.1, EC 2.7.7.6, FLJ42929, KIAA1534oxysterol-binding protein-related protein 5, OBPH1, ORP-5, ORP5oxysterol-binding protein homologue 1, OSBP-related protein 5, oxysterol binding protein-like 5, oxysterol-binding protein homolog 1, Oxysterol-binding protein homolog 1;
Goat
Affinity Purified
RUO
114879
Human
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Western Blot 0.2 - 0.6 ug/mL, Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:32000
NP_065947
OSBPL5
Peptide with sequence KEEAFLRRRFSLC corresponding to N-Terminus according to NP_065947.1, NP_663613.1, NP_001137535.1.
0.1 mg
Primary
This antibody is expected to recognize both reported isoforms. Variants (NP_663613.1; NP_001137535.1) encode the same isoform.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only