missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38107-20ul
This item is not returnable.
View return policy
Description
OSBPL3 Polyclonal antibody specifically detects OSBPL3 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| OSBPL3 | |
| Polyclonal | |
| Western Blot 1:200 - 1:2000, ELISA | |
| DKFZp667P1518, KIAA0704ORP3ORP-3OSBP3, MGC21526, OSBP-related protein 3, oxysterol binding protein-like 3, oxysterol-binding protein 3, oxysterol-binding protein-related protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 370-520 of human OSBPL3 (NP_056365.1).,, Sequence:, SSSPSAQVIGLKNALSSALAQNTDLKERLRRIHAESLLLDSPAVAKSGDNLAEENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPC | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26031 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction