missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL2 Antibody (OTI2E6), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73161PCP
This item is not returnable.
View return policy
Description
OSBPL2 Monoclonal antibody specifically detects OSBPL2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| OSBPL2 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI2E6 | |
| Western Blot | |
| KIAA0772FLJ20223, ORP-2MGC4307, ORP2MGC8342, OSBP-related protein 2, oxysterol binding protein-like 2, oxysterol-binding protein-like 2, oxysterol-binding protein-related protein 2 | |
| Human recombinant protein fragment corresponding to amino acids 258-480 of human OSBPL2 (NP_653081) produced in E.coli. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 9885 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction