missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11318-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.Specifications
| OSBPL11 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL11 | |
| ORP 11, ORP11, OSBP related protein 11, OSBP12, OSBPL11, TCCCIA00292 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 106326, 114885 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CI95, Q9BXB4 | |
| Osbpl11 | |
| OSBPL11 Fusion Protein Ag1855 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction