missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OSBPL11 Monoclonal Antibody (OTI6H9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525293
This item is not returnable.
View return policy
Description
OSBPL11 Monoclonal Antibody for Western Blot, ICC/IF
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
Specifications
| OSBPL11 | |
| Monoclonal | |
| 0.96 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BXB4 | |
| OSBPL11 | |
| Full length human recombinant protein of OSBPL11 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Canine, Monkey | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| OTI6H9 | |
| Unconjugated | |
| OSBPL11 | |
| 9430097N02Rik; AI132306; AU019795; FLJ13012; FLJ13164; ORP11; ORP-11; OSBP12; OSBPL11; OSBP-related protein 11; oxysterol binding protein like 11; oxysterol binding protein-like 11; oxysterol-binding protein-related protein 11; TCCCIA00292; testicular tissue protein Li 132 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100856388, 114885 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction