missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36544
This item is not returnable.
View return policy
Description
OSBPL11 Polyclonal specifically detects OSBPL11 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| OSBPL11 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| FLJ13012, FLJ13164, ORP11OSBP-related protein 11, ORP-11TCCCIA00292, OSBP12, oxysterol binding protein-like 11, oxysterol-binding protein-related protein 11 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1: 10000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_073613.2 | |
| OSBPL11 | |
| The immunogen recognized by the antibody maps to a region between residue 1 to 50 of human Oxysterol Binding Protein-Like 11 using the numbering given in entry NP_073613.2. | |
| 100 μL | |
| Lipid and Metabolism | |
| 114885 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction