Learn More
Invitrogen™ OSBPL10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589070
Description
Immunogen sequence: ASPCSQRHLS VGAPGVVTIT HHKSPAAARR AKSQYSGQLH EVREMMNQVE GQQKNLVHAI ESLPGSGPLT ALDQDLLLLK ATSAATLSCL GECLNLLQQS VHQAGQPSQK PGASENILGW HGSKSHSTEQ LKNGTLGSLP SASANITWAI LPNSAEDEQT SQPEPEPNSG SELVLSEDEK SDNEDKEETE LGVMEDQRSI I; Positive Samples: A-431, HT-29, A-549, Rat brain; Cellular Location: Cytoplasm, cytoskeleton.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
Specifications
| OSBPL10 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL10 | |
| 4933433D06Rik; C820004B04Rik; FLJ20363; HGNC:16395; OPR-10; ORP10; ORP-10; OSBP9; OSBPL10; OSBP-related protein 10; OTTHUMP00000161257; oxysterol binding protein like 10; oxysterol binding protein-like 10; oxysterol-binding protein-related protein 10; RGD1561652 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114884, 316039, 74486 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.38 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BXB5, S4R1M9 | |
| OSBPL10 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human OSBPL10 (NP_0602542). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.