missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSBPL10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15491-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| OSBPL10 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL10 | |
| ORP 10, ORP10, OSBP related protein 10, OSBP9, OSBPL10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114884, 316039, 74486 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BXB5, S4R1M9 | |
| Osbpl10 | |
| OSBPL10 Fusion Protein Ag7861 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction