missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ OSBPL10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29262
This item is not returnable.
View return policy
Description
OSBPL10 Polyclonal antibody specifically detects OSBPL10 in Human samples. It is validated for Immunoprecipitation
Specifications
| OSBPL10 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| ORP-10, ORP10FLJ20363, OSBP9, OSBP-related protein 10, oxysterol binding protein-like 10, oxysterol-binding protein-related protein 10 | |
| Between 1 and 50 | |
| 100 μg | |
| Stem Cell Markers | |
| 114884 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction