missing translation for 'onlineSavingsMsg'
Learn More

γ-Oryzanol, MedChemExpress

Product Code. 16422389
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422389 Supplier Medchem Express Supplier No. HYB2194

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress γ-Oryzanol is a potent DNA methyltransferases (DNMTs) inhibitor in the striatum of mice. γ-Oryzanol significantly inhibits the activities of DNMT1 (IC50=3.2 μM), DNMT3a (IC50=22.3 μM).

γ-Oryzanol is a potent DNA methyltransferases (DNMTs) inhibitor in the striatum of mice. γ-Oryzanol significantly inhibits the activities of DNMT1 (IC50=3.2 μM), DNMT3a (IC50=22.3 μM).

Specifications

Chemical Name or Material γ-Oryzanol
CAS 11042-64-1
Color White
Purity Grade Notes Research
Molecular Formula C40H58O4
Quantity 1 g
Solubility Information DMSO : 62.5 mg/mL (103.67 mM; Need ultrasonic)
SMILES C[C@@H]([C@@]1([H])CC[C@]2(C)[C@]1(C)CCC34C2CCC5[C@@]3(CC[C@H](OC(/C=C/C6=CC(OC)=C(O)C=C6)=O)C5(C)C)C4)CC/C=C(C)\C
Molecular Weight (g/mol) 602.89
Formula Weight 602.89
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.